4-tert-Butylphenylmagnesium bromide 2 M in Diethylether, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

432861000
432861000.EA 281 EUR
432861000.
4-tert-Butylphenylmagnesium bromide 2 M in Diethylether, AcroSeal®
4-tert-Butylphenylmagnesium bromide
Formel: C₁₀H₁₃BrMg
Molecular Weight: 237,42 g/mol
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013244
CAS-Nummer: 63488-10-8
UN: 2924

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Solution
Titration Grignard 1.50 to 2.10 M
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR