(±)-1-Ethoxy-2-propanol ≥90% remainder 2-ethoxy-1-propanol
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether
43306.AP
43306.AY
43306.APEA
106
EUR
43306.AP
43306.AY
(±)-1-Ethoxy-2-propanol ≥90% remainder 2-ethoxy-1-propanol
(±)-1-Ethoxy-2-propanol
Purity: 90+%
CAS No.: 1569-02-4
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 42°(108°F)
Physical Form: Liquid
Density: 0.896
Refractive Index: 1.4065
Appearance: Colourless
Melting Point: -100°
Boiling Point: 132°
UN No.: 1987
MDL No.: MFCD00067050
Formel:
C₅H₁₂O₂ Molecular Weight: 104.15 g/mol Siedepunkt: 132 °C Schmelzpunkt: –100 °C Dichte: 0.896 Flammpunkt: 42 °C (108 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067050 CAS-Nummer: 1569-02-4 EINECS: 216-374-5 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...