(±)-1-Ethoxy-2-propanol ≥90% remainder 2-ethoxy-1-propanol

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether

43306.AP 43306.AY
43306.APEA 106 EUR
43306.AP 43306.AY
(±)-1-Ethoxy-2-propanol ≥90% remainder 2-ethoxy-1-propanol
(±)-1-Ethoxy-2-propanol

Purity: 90+% CAS No.: 1569-02-4 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 42°(108°F) Physical Form: Liquid Density: 0.896 Refractive Index: 1.4065 Appearance: Colourless Melting Point: -100° Boiling Point: 132° UN No.: 1987 MDL No.: MFCD00067050

Formel: C₅H₁₂O₂
Molecular Weight: 104.15 g/mol
Siedepunkt: 132 °C
Schmelzpunkt: –100 °C
Dichte: 0.896
Flammpunkt: 42 °C (108 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00067050
CAS-Nummer: 1569-02-4
EINECS: 216-374-5
UN: 1987
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR