Organic reference standard, Citral, analytical standard, Supelco®
Lieferant: Merck
43318-1ML
43318-1MLEA 52.8 EUR
43318-1ML
Organic reference standard, Citral, analytical standard, Supelco®
Standards Organische Standards
Citral usually is a key component of lemon scented essential oils and also in volatile oils of plants like ginger, which can exist like a mixture of two isomers, cis-isomer neral and trans-isomer geranial. It can serve as potential candidate used in the treatment of tumours. It can mainly find applications as a food additive and as a fragrance in cosmetics.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4afe1350779dc233cb3f2560ebf702a7895b2b6&vid=c4afe1350779dc233cb3f2560ebf702a7895b2b6&prid=web&clid=SAPDAM