Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate

Lieferant: Thermo Scientific
Achtung

43345.02
43345.02EA 236 EUR
43345.02
Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate
Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate

CAS No.: 14054-87-6 Molecular Formula: C24H12F9O6S3Eu.xH2O Formula Weight: 815.51(anhy) Physical Form: Powder Appearance: Yellow Melting Point: 143-144° Solubility: Soluble in methanol, toluene MDL No.: MFCD00041948


Applications: As a laser dye

Formel: C₂₄H₁₇EuF₉O₇S₃
Molecular Weight: 851,818 g/mol
Schmelzpunkt: 143…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041948
CAS-Nummer: 14054-87-6
EINECS: 237-892-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR