Sodium benzyloxide 1 M in Benzylalkohol, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
sodium phenylmethoxide, Sodium Phenylmethanolate
433551000
433551000.EA
216
EUR
433551000.
Sodium benzyloxide 1 M in Benzylalkohol, AcroSeal®
Sodium benzyloxide
Formel:
C₇H₇NaO Siedepunkt: 203…205 °C Dichte: 1,077 g/cm³ Flammpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192444 CAS-Nummer: 20194-18-7 UN: 3267 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Viscous liquid |
Strength | 0.95 to 1.10 M |
Loss on drying | ≤2 % |
Titration with HClO4 | ≥99.0 % (On dry substance) |
Dnase,Rnase,Protease activity | None detected |
Heavy metals (as Pb) | ≤4 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...