2-Pyridylzinc bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

433630500
433630500.EA 183 EUR
433630500.
2-Pyridylzinc bromide 0.5 M in THF, AcroSeal®
2-Pyridylzinc bromide
Formel: C₅H₄BrNZn
Dichte: 0,974 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00671985
CAS-Nummer: 218777-23-2
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to brown to black Solution
Titration Argentometric 0.45 to 0.65 M
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR