Gold(I)chlorid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

43365.03 43365.06
43365.03EA 264 EUR
43365.03 43365.06
Gold(I)chlorid ≥99,9% (Metall-Basis)
Gold(I)chlorid

Purity: 99.9% CAS No.: 10294-29-8 Molecular Formula: AuCl Formula Weight: 232.42 Physical Form: Powder Density: 7.57 Melting Point: 289° dec. Merck Reference: 14,4515 UN No.: 3260 MDL No.: MFCD00046175

Formel: AuCl
Dichte: 7,57
MDL: MFCD00046175
CAS-Nummer: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8,III
Merck Index: 14,4515

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR