Lithiumwolframat ≥99%

Lieferant: Thermo Fisher Scientific

43366.30 43366.18
43366.30EA 429 EUR
43366.30 43366.18
Lithiumwolframat ≥99%
Lithiumwolframat

Purity: 99% min CAS No.: 13568-45-1 Molecular Formula: Li2WO4 Formula Weight: 261.73 Physical Form: Powder Density: 3.71 Melting Point: 742° MDL No.: MFCD00016185

Formel: Li₂O₄W
Schmelzpunkt: 742 °C
Dichte: 3,71
Lagertemperatur: Raumtemperatur
MDL: MFCD00016185
CAS-Nummer: 13568-45-1
EINECS: 236-978-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR