4-Methyl-1-naphthylmagnesium bromide 0.25 M in THF, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
433690500
433690500.EA
126
EUR
433690500.
4-Methyl-1-naphthylmagnesium bromide 0.25 M in THF, AcroSeal®
4-Methyl-1-naphthylmagnesium bromide
Formel:
C₁₁H₉BrMg Molecular Weight: 245,402 g/mol Dichte: 0,948 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311487 CAS-Nummer: 115913-58-1 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Yellow to brown Solution |
Titration Grignard | 0.22 to 0.28 M |
Water | ≤3.0 % |
pH | 6.0 to 8.0 (10 % in water) |
Hydroxyl value | 65 to 78 mg KOH/g |
Iodine value | 25 to 35 g I2/100 g |
Saponification value | 65 to 70 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...