N,N,N',N'-Tetramethylethylendiamin (TEMED) 99.5%, AcroSeal®, purified_redistillation
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,2-Bis(dimethylamino)ethan
433831000
433831000.EA
85.8
EUR
433831000.
N,N,N',N'-Tetramethylethylendiamin (TEMED) 99.5%, AcroSeal®, purified_redistillation
N,N,N',N'-Tetramethylethylendiamin (TEMED)
Formel:
C₆H₁₆N₂ Molecular Weight: 116.21 g/mol Siedepunkt: 120…122 °C (760 mmHg) Schmelzpunkt: –55 °C Dichte: 0.77 g/cm³ Flammpunkt: 17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008335 CAS-Nummer: 110-18-9 UN: 2372 ADR: 3 II |
|
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4190 (20°C, 589 nm) |
GC | ≥99.4 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...