Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 4-aminothiophene-3-carboxylate hydrochloride
43384.03
43384.06
43384.03EA
83.3
EUR
43384.03
43384.06
Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%
Methyl-4-aminothiophene-3-carboxylate hydrochloride
Purity: 97+%
CAS No.: 39978-14-8
Molecular Formula: C6H7NO2S.HCl
Formula Weight: 193.65
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Brown
Melting Point: 199-200° dec.
MDL No.: MFCD00068149
Formel:
C₆H₈ClNO₂S Schmelzpunkt: 199…200 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068149 CAS-Nummer: 39978-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...