Tantaltris(diethylamido)-tert-butylimid ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
43419.14
43419.06
43419.14EA
863
EUR
43419.14
43419.06
Tantaltris(diethylamido)-tert-butylimid ≥99,99% (Metall-Basis)
Tantaltris(diethylamido)-tert-butylimid
Purity: 99.99%
CAS No.: 169896-41-7
Molecular Formula: C16H39N4Ta
Formula Weight: 468.48
Physical Form: Liquid, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Yellow
Boiling Point: 90°/0.1mm
UN No.: 3129
MDL No.: MFCD02684506
Precursor for TaN
Formel:
C₁₆H₃₉N₄Ta Siedepunkt: 90 °C (0,1 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684506 CAS-Nummer: 169896-41-7 UN: 3129 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...