Tantaltris(diethylamido)-tert-butylimid ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

43419.14 43419.06
43419.14EA 863 EUR
43419.14 43419.06
Tantaltris(diethylamido)-tert-butylimid ≥99,99% (Metall-Basis)
Tantaltris(diethylamido)-tert-butylimid

Purity: 99.99% CAS No.: 169896-41-7 Molecular Formula: C16H39N4Ta Formula Weight: 468.48 Physical Form: Liquid, Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Yellow Boiling Point: 90°/0.1mm UN No.: 3129 MDL No.: MFCD02684506


Precursor for TaN

Formel: C₁₆H₃₉N₄Ta
Siedepunkt: 90 °C (0,1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02684506
CAS-Nummer: 169896-41-7
UN: 3129
ADR: 4.3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR