1-Butanthiol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: n-Butylmercaptan, 1-Mercaptobutan, NBM, n-Butanthiol

43421.AE 43421.AP 43421.AC
43421.AEEA 48.2 EUR
43421.AE 43421.AP 43421.AC
1-Butanthiol ≥98%
1-Butanthiol

Purity: 98% CAS No.: 109-79-5 Molecular Formula: C4H10S Formula Weight: 90.19 Flash Point: 2°(36°F) Physical Form: Liquid Density: 0.842 Refractive Index: 1.4425 Sensitivity: Air Sensitive Appearance: Colourless Odour: Stench Melting Point: -116° Boiling Point: 98° Beilstein Reference: 1730908 Merck Reference: 14,1577 UN No.: 2347 MDL No.: MFCD00004905

Formel: C₄H₁₀S
Molecular Weight: 90.19 g/mol
Siedepunkt: 98 °C
Schmelzpunkt: –116 °C
Dichte: 0.842
Flammpunkt: 2 °C (36 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004905
CAS-Nummer: 109-79-5
EINECS: 203-705-3
UN: 2347
ADR: 3 II
Merck Index: 12,01611

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR