Zink(II)nitrat Hexahydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Zinknitrat Hexahydrat

4344-01EA 107.7 EUR
4344-01 4344-05
Zink(II)nitrat Hexahydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Zink(II)nitrat Hexahydrat
Formel: ZnN₂O₆·6H₂O
Siedepunkt: ∼105 °C (1013 hPa)
Schmelzpunkt: 36 °C
Dichte: 2,06 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149889
CAS-Nummer: 10196-18-6
EINECS: 231-943-8
UN: 1514
ADR: 5.1,II
Merck Index: 12,10273

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 99.0 - 101.0 %
IMPORTANT: Store below 24°C (75°F). Ship in refrigerated trucks only. = 0.005 %
Assay (Zn(NO₃)₂·6H₂O) (by EDTA titrn) 99.0 - 101.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 3.5 - 5.5
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Lead (Pb) ≤0.005 %
Trace Impurities - Copper (Cu) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR