Nickel(II)chlorid, wasserfrei ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Nickeldichlorid

43441.14 43441.06
43441.14EA 646 EUR
43441.14 43441.06
Nickel(II)chlorid, wasserfrei ≥99,99% (Metall-Basis)
Nickel(II)chlorid

Purity: 99.99% CAS No.: 7718-54-9 Molecular Formula: Cl2Ni Formula Weight: 129.60 Physical Form: Crystalline powder Density: 3.55 Appearance: Yellow Odour: Odourless Melting Point: 1001° Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00011142

Formel: Cl₂Ni
Molecular Weight: 129.6 g/mol
Schmelzpunkt: 1001 °C
Dichte: 3.55
Lagertemperatur: Raumtemperatur
MDL: MFCD00011142
CAS-Nummer: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,6505

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR