Polyethylenglycol 8.000
Lieferant: Thermo Fisher Scientific
43443.A3
43443.22
43443.A9
43443.36
43443.A3EA
131
EUR
43443.A3
43443.22
43443.A9
43443.36
Polyethylenglycol 8.000
Polyethylenglycol 8.000
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Physical Form: Powder
Appearance: White
Odour: Odourless
pH: 5-7
Melting Point: 55-63°
Merck Reference: 14,7568
MDL No.: MFCD01779615
Formel:
H(OCH₂CH₂)nOH Siedepunkt: 250 °C (1013 hPa) Schmelzpunkt: 55…63 °C Dichte: 1,125 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01779615 CAS-Nummer: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...