Natriumiodid, ultra trocken ≥99,98% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
43447.22
43447.14
43447.36
43447.22EA
316
EUR
43447.22
43447.14
43447.36
Natriumiodid, ultra trocken ≥99,98% (Metall-Basis)
Natriumiodid
Purity: 99.98%
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.67
Refractive Index: 1.7745
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 651°
Boiling Point: 1300-1304°
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formel:
NaI Siedepunkt: 1300…1304 °C Schmelzpunkt: 651 °C Dichte: 3,67 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003532 CAS-Nummer: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9,III Merck Index: 14,8631 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...