Eisen(II)bromid, ultra trocken ≥99,995% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Eisendibromid

Unter Argon in Ampullen verpackt

43472.14
43472.14EA 480 EUR
43472.14
Eisen(II)bromid, ultra trocken ≥99,995% (Metall-Basis)
Eisen(II)bromid

Purity: 99.995% CAS No.: 7789-46-0 Molecular Formula: Br2Fe Formula Weight: 215.67 Physical Form: Powder, Ampouled under argon Density: 4.636 Melting Point: 684° Merck Reference: 14,4042 UN No.: 3260 MDL No.: MFCD00016081

Formel: Br₂Fe
Molecular Weight: 215,66 g/mol
Schmelzpunkt: 684 °C
Dichte: 4,636
Lagertemperatur: Raumtemperatur
MDL: MFCD00016081
CAS-Nummer: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8,III
Merck Index: 14,4042

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR