Ammonium tetrathiomolybdate ≥99,95% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
43493.14
43493.06
43493.14EA
548
EUR
43493.14
43493.06
Ammonium tetrathiomolybdate ≥99,95% (Metall-Basis)
Ammonium tetrathiomolybdate
Purity: 99.95%
CAS No.: 15060-55-6
Molecular Formula: H8MoN2S4
Formula Weight: 260.27
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Black
MDL No.: MFCD00136013
Formel:
H₁₀MoN₂S₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136013 CAS-Nummer: 15060-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...