Etidronisäure ≥96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: HEDP, OEDFK, 1-Hydroxyethylidenediphosphonic acid, 1-Hydroxyethane-1,1-diphosphonic Acid, HDEPA, 1-Hydroxyethylidene-1,1-diphosphonic acid, 1-Hydroxyethylidenebis(phosphonic acid)

43502.C4
43502.25EA 0 EUR
43502.25
Etidronisäure ≥96%
Etidronisäure

Purity: 96% CAS No.: 2809-21-4 Molecular Formula: CH3C(OH)(PO3H2)2 Formula Weight: 206.03 Physical Form: Powder Odour: Odourless Merck Reference: 14,3863 UN No.: 3261 MDL No.: MFCD00070585

Formel: C₂H₈O₇P₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00070585
CAS-Nummer: 2809-21-4
EINECS: 220-552-8
UN: 3261
ADR: 8,III
Merck Index: 14,3863

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR