Zinkoxid ≥99,0%, Pulver, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Zink(II)oxid

4358-05EA 345.16 EUR
4358-05 4358-01 4358-R
Zinkoxid ≥99,0%, Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Zinkoxid
Formel: ZnO
Molecular Weight: 81.39 g/mol
Schmelzpunkt: 1975 °C
Dichte: 5.61 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011300
CAS-Nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Merck Index: 13,10200

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.01%
ACS - Assay (ZnO) ≥99.0 %
ACS - Insoluble in Dilute H₂SO₄ ≤0.01 %
ACS - Alkalinity Passes Test
ACS - Nitrate (NO₃) ≤0.003 %
ACS - Sulfur Compounds (as SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Lead (Pb) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤5.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR