Pentadecylmagnesium bromide 0.5 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
436160500
436160500.EA
268
EUR
436160500.
Pentadecylmagnesium bromide 0.5 M in THF, AcroSeal®
Pentadecylmagnesium bromide
Formel:
C₁₅H₃₁BrMg Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209607 CAS-Nummer: 78887-70-4 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Yellow to brown Solution |
Titration Grignard | 0.45 to 0.55 M |
Melting point | 80 to 83 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...