4-Benzyloxyphenylmagnesium bromide 0.8 M in THF, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
[4-(Benzyloxy)phenyl](bromo)magnesium, Magnesium, bromo[4-(phenylmethoxy)phenyl]-
436221000
436221000.EA
240
EUR
436221000.
4-Benzyloxyphenylmagnesium bromide 0.8 M in THF, AcroSeal®
4-Benzyloxyphenylmagnesium bromide
Formel:
C₁₃H₁₁BrMgO Molecular Weight: 287,439 g/mol Dichte: 1,033 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09039130 CAS-Nummer: 120186-59-6 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to brown Solution |
Titration Grignard | 0.70 to 1.05 M |
Melting point | 56 to 60 °C |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...