Agarose, Electran, resolution_high, low melting Electrophoresis

Lieferant: VWR Chemicals

436552VEA 405 EUR
436552V 436553W
Agarose, Electran, resolution_high, low melting Electrophoresis
Agarose

Agarose high resolution is optimised to give exceptional separation on agarose of nucleic acid fragments below 1000 base pairs, and differing by only a few base pairs. It retains the low gelling characteristics of the standard Agarose HR, and offers significantly increased gel strength at the expense of a higher melting temperature. This results in gels which are flexible and easy to handle. These properties make it the agarose of choice for analytical gel separations to check the quality and size of amplified fragments, and for use in restriction mapping involving small digestion fragments.


  • Free from DNases and RNases

Schmelzpunkt: 60…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081294
CAS-Nummer: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Moisture content Max. 10 %
Sulfate content Max. 0.15 %
Gelstärke (4%) Min. 500 g/cm²
EEO, -Mr Max. 0.15
Geliertemperature (4%) Max. 35 °C
Schmelztemperature (4%) Max. 65 °C
Appearance Passes
Ash content Max. 1 %
DNA binding Passes
RNase / DNase Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR