Supelco® Organic Reference Standard, Dopamine Hydrochloride
Lieferant: Merck
43658-50MG
43658-50MGEA 182 EUR
43658-50MG
Supelco® Organic Reference Standard, Dopamine Hydrochloride
Standards Organische Standards
  • GHS signal: GHS07, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P261 - P272 - P280 - P302 + P352 - P333 + P313 - P362 + P364
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f20984f0bac15a5b7fd75ef9f048a339b3d8061c&vid=3952b8cbbadd9072fc893585b4731f97c74d9ff8&prid=web&clid=SAPDAM