Calcium molybdate ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

43686.22 43686.14
43686.22EA 113 EUR
43686.22 43686.14
Calcium molybdate ≥99%
Calcium molybdate

Purity: 99% CAS No.: 7789-82-4 Molecular Formula: CaMoO4 Formula Weight: 200.01 Physical Form: Powder Density: 4.45 Appearance: White Odour: Odourless Melting Point: 965° MDL No.: MFCD00015976

Formel: CaMoO₄
Schmelzpunkt: 965 °C
Dichte: 4,45
Lagertemperatur: Raumtemperatur
MDL: MFCD00015976
CAS-Nummer: 7789-82-4
EINECS: 232-192-9
Merck Index: 14,1681

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR