Palladium(II)sulfat Dihydrat ≥99,995% (Metall-Basis), Premion®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
43698.03
43698.06
43698.03EA
189
EUR
43698.03
43698.06
Palladium(II)sulfat Dihydrat ≥99,995% (Metall-Basis), Premion®
Palladium(II)sulfat Dihydrat
Purity: 99.995%
CAS No.: 13444-98-9
Molecular Formula: O4PdS.2H2O
Formula Weight: 238.51 (202.48anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Red to brown
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00011173
Formel:
O₄PdS·2H₂O Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011173 CAS-Nummer: 13444-98-9 EINECS: 236-957-8 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...