Niob(V)chlorid ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Niobpentachlorid

unter Argon abgefüllt

43729.22 43729.14 43729.06
43729.22EA 1198 EUR
43729.22 43729.14 43729.06
Niob(V)chlorid ≥99,999% (Metall-Basis), Puratronic®
Niob(V)chlorid

Purity: 99.999% CAS No.: 10026-12-7 Molecular Formula: NbCl5 Formula Weight: 270.20 Physical Form: Powder, Packaged under argon Density: 2.74 Melting Point: 208.3° Boiling Point: 248.2° Merck Reference: 14,6557 UN No.: 3260 MDL No.: MFCD00011127

Formel: Cl₅Nb
Molecular Weight: 270.17 g/mol
Siedepunkt: 248.2 °C
Schmelzpunkt: 208.3 °C
Dichte: 2.74
Lagertemperatur: Raumtemperatur
MDL: MFCD00011127
CAS-Nummer: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8 III
Merck Index: 14,6557

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR