2,1,3-Benzothiadiazol ≥98%

Lieferant: Thermo Scientific

43730.09
43730.09EA 77.7 EUR
43730.09
2,1,3-Benzothiadiazol ≥98%
2,1,3-Benzothiadiazole

Purity: 98% CAS No.: 273-13-2 Molecular Formula: C6H4N2S Formula Weight: 136.18 Flash Point: 95°(203°F) Physical Form: Powder Melting Point: 42-44° Boiling Point: 206° MDL No.: MFCD00005809

Formel: C₆H₄N₂S
Molecular Weight: 136,18 g/mol
Siedepunkt: 206 °C
Schmelzpunkt: 42…44 °C
Flammpunkt: 95 °C (203 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005809
CAS-Nummer: 273-13-2
EINECS: 205-985-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR