Tantalpentachlorid, wasserfrei ≥99,995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Tantal(V)chlorid

Unter Argon in Ampullen verpackt

43731.22 43731.06
43731.22EA 489 EUR
43731.22 43731.06
Tantalpentachlorid, wasserfrei ≥99,995% (Metall-Basis), Puratronic®
Tantalpentachlorid

Purity: 99.995% CAS No.: 7721-01-9 Molecular Formula: TaCl5 Formula Weight: 358.21 Physical Form: Powder, Ampouled under argon Density: 3.68 Appearance: White to pale yellow Odour: Acidic Melting Point: 216° Boiling Point: 242° Merck Reference: 14,9054 UN No.: 3260 MDL No.: MFCD00011253

Formel: Cl₅Ta
Siedepunkt: 242 °C
Schmelzpunkt: 216 °C
Dichte: 3,68
Lagertemperatur: Raumtemperatur
MDL: MFCD00011253
CAS-Nummer: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III
Merck Index: 14,9054

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR