Leucomalachitgrün
Lieferant: Thermo Fisher Scientific
43758.09
43758.04
43758.09EA
109
EUR
43758.09
43758.04
Leucomalachitgrün
Leuco malachite green
CAS No.: 129-73-7
Molecular Formula: C23H26N2
Formula Weight: 330.47
Physical Form: Powder
Sensitivity: Air Sensitive
Melting Point: 100-102°
Beilstein Reference: 2140506
MDL No.: MFCD00008315
Formel:
C₂₃H₂₆N₂ Schmelzpunkt: 100…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008315 CAS-Nummer: 129-73-7 EINECS: 204-961-9 Merck Index: 14,5699 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...