Propargylamin Hydrochlorid ≥95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Propinylammoniumchlorid, Propargylammoniumchlorid, Monopropargylamin Hydrochlorid, Prop-2-ynylamin Hydrochlorid, 3-Aminopropin Hydrochlorid

43775.03 43775.06
43775.03EA 33.5 EUR
43775.03 43775.06
Propargylamin Hydrochlorid ≥95%
Propargylamin Hydrochlorid

Purity: 95% CAS No.: 15430-52-1 Molecular Formula: HC≡CCH2NH2.HCl Formula Weight: 91.54 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 2811 MDL No.: MFCD00012907

Formel: C₃H₆ClN
Schmelzpunkt: 179…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012907
CAS-Nummer: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR