Ethyl-2-cyanoacetoacetate
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
43792.09
43792.04
43792.09EA
146
EUR
43792.09
43792.04
Ethyl-2-cyanoacetoacetate
Ethyl-2-cyanoacetoacetate
CAS No.: 634-55-9
Molecular Formula: C2H5OOCCH(CN)COCH3
Formula Weight: 155.16
Physical Form: Liquid
UN No.: UN3276
MDL No.: MFCD00043487
Formel:
C₇H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043487 CAS-Nummer: 634-55-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...