Zinksulfat Heptahydrat ≥99%, körnig USP, Multikompendial, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Zinkvitriol

4383-01EA 125.74 EUR
4383-01
Zinksulfat Heptahydrat ≥99%, körnig USP, Multikompendial, J.T.Baker®
Zinksulfat Heptahydrat
Formel: ZnSO₄·7H₂O
Molecular Weight: 287,54 g/mol
Schmelzpunkt: 100 °C
Dichte: 1,97 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149894
CAS-Nummer: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity Passes Test
USP - Alkalies and Alkaline Earths ≤0.9 %
USP - Arsenic (As) ≤14 ppm
USP - Lead (Pb) ≤0.002 %
USP - Assay (ZnSO₄·7H₂O) 99.0 - 108.7 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 4.4 - 5.6
EP/BP - Chloride (Cl) ≤300 ppm
EP/BP - Iron (Fe) ≤100 ppm
EP/BP - Assay (ZnSO₄·7H₂O) (dried basis) 99.0 - 104.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR