Iridium ≥99,95% (Metall-Basis), Perlen 6-7 mm (0.24-0.28 in)

Lieferant: Thermo Scientific

43859.KG
43859.KGEA 3140 EUR
43859.KG
Iridium ≥99,95% (Metall-Basis), Perlen 6-7 mm (0.24-0.28 in)
Iridium

Purity: 99.95% CAS No.: 7439-88-5 Physical Form: ≈4.5g/piece MDL No.: MFCD00011062

Formel: Ir
Molecular Weight: 192,22 g/mol
Siedepunkt: 5300 °C (1013 hPa)
Schmelzpunkt: 2450 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011062
CAS-Nummer: 7439-88-5
EINECS: 231-095-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR