Aluminum oxide-Silicon oxide 13%, kleine Fläche makroporöser Katalysatorträger
Lieferant: Thermo Fisher Scientific
43863.A1
43863.A7
43863.A1EA
240
EUR
43863.A1
43863.A7
Aluminum oxide-Silicon oxide 13%, kleine Fläche makroporöser Katalysatorträger
Aluminum oxide-Silicon oxide
Physical Form: 3/16" Spheres
MDL No.: MFCD00058866
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...