Pentafluorophenylmagnesium bromide 0.5 M in Diethylether, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

438741000
438741000.EA 181 EUR
438741000.
Pentafluorophenylmagnesium bromide 0.5 M in Diethylether, AcroSeal®
Pentafluorophenylmagnesium bromide
Formel: C₆BrF₅Mg
Dichte: 0,86 g/cm³
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015720
CAS-Nummer: 879-05-0
UN: 2924
ADR: 3,I

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown to black Liquid
Titration Grignard 0.4 to 0.6 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR