Tetrahexylammoniumhydrogensulfat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

43882.04
43882.04EA 133 EUR
43882.04
Tetrahexylammoniumhydrogensulfat ≥98%
Tetrahexylammonium hydrogen sulphate

Purity: 98% CAS No.: 32503-34-7 Molecular Formula: (C6H13)4NHSO4 Formula Weight: 451.76 Physical Form: Powder Melting Point: 96-102° MDL No.: MFCD00037675

Formel: C₂₄H₅₃NO₄S
Molecular Weight: 451,76 g/mol
Schmelzpunkt: 96…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037675
CAS-Nummer: 32503-34-7
EINECS: 251-069-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR