(Aminoethylaminomethyl)phenethyltrimethoxysilanes Gemisch aus m- und p-Isomeren

Lieferant: Thermo Scientific
Gefahr

43904.04 43904.09 43904.18
43904.04EA 37.6 EUR
43904.04 43904.09 43904.18
(Aminoethylaminomethyl)phenethyltrimethoxysilanes Gemisch aus m- und p-Isomeren
(Aminoethylaminomethyl)phenethyltrimethoxysilanes

CAS No.: 74113-77-2 Molecular Formula: H2NC2H4NHCH2C6H4C2H4Si(OCH3)3 Formula Weight: 298.46 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.02 Sensitivity: Air and Moisture Sensitive Appearance: Colourless to pale yellow Boiling Point: 128°/0.2mm UN No.: UN2735 MDL No.: MFCD00054775


Applications: As a crosslinking agent

Formel: H₂NC₂H₄NHCH₂C₆H₄C₂H₄Si(OCH₃)₃
Siedepunkt: 128 °C (0,2 mmHg)
Dichte: 1,02
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00054775
CAS-Nummer: 74113-77-2
UN: 2735
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR