Bismut(III)isopropoxid

Lieferant: Thermo Scientific
Achtung


unter Argon abgefüllt

43938.06
43938.06EA 568 EUR
43938.06
Bismut(III)isopropoxid
Bismut(III)isopropoxid

CAS No.: 15049-67-9 Molecular Formula: C9H21BiO3 Formula Weight: 386.24 Physical Form: Powder, Packaged under argon Sensitivity: Moisture Sensitive Appearance: Dark grey UN No.: 3181 MDL No.: MFCD00799065

Formel: C₉H₂₁BiO₃
Molecular Weight: 386,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00799065
CAS-Nummer: 15049-67-9
UN: 3181
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR