Silver protein strong, powder Histology

Lieferant: Thermo Fisher Scientific

Synonyme: Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver

43970.06 43970.14 43970.22
43970.06EA 35.1 EUR
43970.06 43970.14 43970.22
Silver protein strong, powder Histology
Silver protein

Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.


  • Silver content: min 7.5%
  • Hygroscopic
  • Light Sensitive
  • Store under Nitrogen
  • Soluble in water. Slightly soluble in methanol, diethyl ether


CAS No.: 9008-42-8 Physical Form: Powder Sensitivity: Light Sensitive; Hygroscopic Appearance: Pale yellow to brown MDL No.: MFCD00132268


Applications: Histological stain for microscopic analysis

Formel: Ag (with protein)
MDL: MFCD00132268
CAS-Nummer: 9008-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR