Silver protein strong, powder Histology
Lieferant: Thermo Fisher Scientific
Synonyme:
Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver
Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.
- Silver content: min 7.5%
- Hygroscopic
- Light Sensitive
- Store under Nitrogen
- Soluble in water. Slightly soluble in methanol, diethyl ether
CAS No.: 9008-42-8
Physical Form: Powder
Sensitivity: Light Sensitive; Hygroscopic
Appearance: Pale yellow to brown
MDL No.: MFCD00132268
Applications: Histological stain for microscopic analysis
Formel:
Ag (with protein) |
MDL:
MFCD00132268 CAS-Nummer: 9008-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...