Magnesiumoxalat Dihydrat ≥98,5%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
43976.22
43976.36
43976.A3
43976.22EA
110
EUR
43976.22
43976.36
43976.A3
Magnesiumoxalat Dihydrat ≥98,5%
Magnesiumoxalat Dihydrat
Purity: 98.5+%
CAS No.: 547-66-0
Molecular Formula: MgC2O4.2H2O
Formula Weight: 148.36 (112.32anhy)
Physical Form: Powder/Pieces
Odour: Odourless
Merck Reference: 14,5676
UN No.: 3288
MDL No.: MFCD00287279
Formel:
C₂MgO₄·2H₂O Schmelzpunkt: 150 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00287279 CAS-Nummer: 547-66-0 EINECS: 208-932-1 ADR: 6.1,III Merck Index: 14,5676 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...