C-276 alloy, Folie, Dicke 0.2 mm (0.008 in)
Lieferant: Thermo Scientific
|
Gefahr
|
43994.GH
43994.GHEA
114
EUR
43994.GH
C-276 alloy, Folie, Dicke 0.2 mm (0.008 in)
C-276 alloy
High strength, corrosion resistant alloy
Ni:Mo:Cr:Fe:W;Co:Mn; 57.5:15.5:15.5:6:3.5:1.5:0.5 approximate wt%