Luteolin, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyron, Natural Yellow 2, 3',4',5,7-Tetrahydroxyflavon
440025-5
440025-5EA
101
EUR
440025-5
Luteolin, Millipore®
Luteolin
Formel:
C₁₅H₁₀O₆ Molecular Weight: 286.24 g/mol |
MDL:
MFCD00017309 CAS-Nummer: 491-70-3 EINECS: 207-741-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...