Matrix oil, hydrocarbon oil, 20 cSt

Lieferant: Thermo Fisher Scientific

44012.X2 44012.XC
44012.X2EA 45.8 EUR
44012.X2 44012.XC
Matrix oil, hydrocarbon oil, 20 cSt
Matrix oil, hydrocarbon oil

Physical Form: Liquid
Odor: Petroleum-like


Physical Form: Liquid MDL No.: MFCD00147539

Lagertemperatur: Raumtemperatur
MDL: MFCD00147539
CAS-Nummer: 64742-65-0
EINECS: 265-169-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR