Blei ≥99,9% (Metall-Basis), Drahtform, Ø 2.0 mm (0.08 in)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44016.G6
44016.H2
44016.G6EA
158
EUR
44016.G6
44016.H2
Blei ≥99,9% (Metall-Basis), Drahtform, Ø 2.0 mm (0.08 in)
Lead
Purity: 99.9%
CAS No.: 7439-92-1
Physical Form: ≈35.6g/m
Appearance: Grey
Odour: Odourless
MDL No.: MFCD00134050
Formel:
Pb Siedepunkt: 1740 °C (1013 hPa) Schmelzpunkt: 327 °C Dichte: 11,3437 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134050 CAS-Nummer: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...