2-Benzyloxyphenylmagnesium bromide 1 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
440320500
440320500.EA
201
EUR
440320500.
2-Benzyloxyphenylmagnesium bromide 1 M in THF, AcroSeal®
2-Benzyloxyphenylmagnesium bromide
Formel:
C₁₃H₁₁BrMgO Dichte: 1,047 g/cm³ Flammpunkt: –20 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08705262 CAS-Nummer: 328000-16-4 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear to hazy yellow to brown Solution |
Titration Grignard | 0.95 to 1.10 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...